siRNA Details
Gene Symbol: KIAA0825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006714588.2 | XP_006714651.1 | 14, 15 | 2762 | ||
XM_011543324.2 | XP_011541626.1 | 14, 15 | 2762 | |||
XM_011543325.2 | XP_011541627.1 | 15, 16 | 2828 | |||
XM_011543327.2 | XP_011541629.1 | 14, 15 | 2678 | |||
XM_011543328.2 | XP_011541630.1 | 15, 16 | 2883 | |||
XM_011543329.2 | XP_011541631.1 | 13, 14 | 2598 | |||
XM_011543330.2 | XP_011541632.1 | 14, 15 | 2762 | |||
XM_011543331.1 | XP_011541633.1 | 14, 15 | 2762 | |||
XM_011543332.1 | XP_011541634.1 | 14, 15 | 2762 | |||
XM_011543333.2 | XP_011541635.1 | 14, 15 | 2762 | |||
XM_011543334.2 | XP_011541636.1 | 14, 15 | 2762 | |||
XM_011543335.2 | XP_011541637.1 | 11, 12 | 2295 | |||
XM_017009373.1 | XP_016864862.1 | 14, 15 | 2762 | |||
XM_017009374.1 | XP_016864863.1 | 14, 15 | 2762 | |||
XM_017009375.1 | XP_016864864.1 | 15, 16 | 2827 | |||
GenBank mRNA | AB020632.1 | - | 8, 9 | 1352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
