siRNA Details
Gene Symbol: IGHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090461.1 | - | 7 | 2044 | ||
AK126280.1 | - | 8 | 1395 | |||
AK128568.1 | - | 9 | 1411 | |||
AK128580.1 | - | 8 | 1395 | |||
AK128614.1 | - | 8 | 1452 | |||
AK128625.1 | - | 8 | 1462 | |||
AK128635.1 | - | 8 | 1473 | |||
AK128649.1 | - | 6 | 1590 | |||
AK128658.1 | - | 7 | 1471 | |||
BC021276.2 | - | 8 | 1421 | |||
BC063384.1 | - | 8 | 1382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
