siRNA Details
Gene Symbol: IGHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090461.1 | - | 3 | 1276 | ||
AK126280.1 | - | 4 | 627 | |||
AK128568.1 | - | 5 | 645 | |||
AK128580.1 | - | 4 | 627 | |||
AK128614.1 | - | 4 | 684 | |||
AK128625.1 | - | 4 | 694 | |||
AK128635.1 | - | 4 | 705 | |||
AK128649.1 | - | 3 | 612 | |||
AK128658.1 | - | 3 | 703 | |||
BC021276.2 | - | 4 | 653 | |||
BC063384.1 | - | 4 | 614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
