siRNA Details
Gene Symbol: PNMAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020709.2 | NP_065760.1 | 1 | 1510 | ||
GenBank mRNA | AB033009.1 | - | 1 | 187 | ||
BC121810.1 | - | 1 | 144 | |||
BC122520.1 | - | 1 | 144 | |||
BX390603.1 | - | 1 | 719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
