siRNA Details
Gene Symbol: PNMAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020709.2 | NP_065760.1 | 1 | 1874 | ||
GenBank mRNA | AB033009.1 | - | 1 | 551 | ||
BC121810.1 | - | 1 | 508 | |||
BC122520.1 | - | 1, 2 | 508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
