siRNA Details
Gene Symbol: TMEM252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153237.1 | NP_694969.1 | 2 | 931 | ||
GenBank mRNA | BC029780.1 | - | 2 | 931 | 5185747 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
