siRNA Details
Gene Symbol: C5orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131091.1 | 4 | 337 | |||
NR_131092.1 | 3 | 253 | ||||
GenBank mRNA | BU561519.1 | - | 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
