siRNA Details
Gene Symbol: DDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015086.1 | NP_055901.2 | 2 | 2262 | ||
XM_011538055.2 | XP_011536357.1 | 3 | 2867 | |||
GenBank mRNA | AB018292.1 | - | 2 | 2146 | ||
BC136398.1 | - | 2 | 2172 | 9020385 | ||
BC136399.1 | - | 2 | 2172 | 9020386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
