siRNA Details
Gene Symbol: GPR26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153442.3 | NP_703143.1 | 3 | 9693 | ||
GenBank mRNA | AK091093.1 | - | 1 | 1601 | ||
AL713636.1 | - | 1 | 1017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
