siRNA Details
Gene Symbol: ZNF852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287349.1 | NP_001274278.1 | 4 | 1609 | ||
GenBank mRNA | AK296954.1 | - | 4 | 1605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
