siRNA Details
Gene Symbol: LINC00670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034144.1 | 2 | 322 | |||
NR_034145.1 | 3 | 484 | ||||
GenBank mRNA | AK092009.1 | - | 3 | 483 | ||
BC122562.1 | - | 2 | 309 | |||
BC122870.1 | - | 2 | 309 | |||
DB255288.1 | - | 3 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
