siRNA Details
Gene Symbol: ANKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144708.3 | NP_653309.3 | 23 | 4339 | ||
XM_011510673.2 | XP_011508975.1 | 23 | 4858 | |||
XM_011510674.1 | XP_011508976.1 | 23 | 4825 | |||
XM_011510675.2 | XP_011508977.1 | 22 | 4753 | |||
XM_011510679.2 | XP_011508981.1 | 21 | 4429 | |||
XM_011510680.1 | XP_011508982.1 | 22 | 4658 | |||
XM_011510685.2 | XP_011508987.1 | 23 | 4414 | |||
XM_011510686.2 | XP_011508988.1 | 19 | 4351 | |||
XM_011510687.2 | XP_011508989.1 | 22 | 4222 | |||
XM_011510691.2 | XP_011508993.1 | 19 | 3044 | |||
XM_011510692.2 | XP_011508994.1 | 21 | 4058 | |||
XM_011510693.2 | XP_011508995.1 | 20 | 3954 | |||
XM_011510694.2 | XP_011508996.1 | 19 | 3773 | |||
XM_017003415.1 | XP_016858904.1 | 18 | 3553 | |||
XM_017003416.1 | XP_016858905.1 | 20 | 3965 | |||
XM_017003418.1 | XP_016858907.1 | 18 | 3668 | |||
XM_017003419.1 | XP_016858908.1 | 18 | 3635 | |||
XM_017003420.1 | XP_016858909.1 | 18 | 3571 | |||
GenBank mRNA | AJ549812.1 | - | 22 | 4038 | ||
AJ605761.1 | - | 10 | 1307 | |||
AK058144.1 | - | 11 | 1691 | |||
AK126073.1 | - | 20 | 3909 | |||
BC037215.1 | - | 16 | 3128 | |||
BC047413.1 | - | 26 | 3903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
