siRNA Details
Gene Symbol: NPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037738.2 | NP_001032827.1 | 9, 10 | 1013 | ||
XM_005265920.3 | XP_005265977.1 | 8, 9 | 982 | |||
GenBank mRNA | AB042278.1 | - | 9, 10 | 768 | ||
AK000472.1 | - | 7, 8 | 576 | |||
BC107754.1 | - | 9, 10 | 869 | 6736004 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001252261, AK184065, BC090843, AK005420, AK088923.
Gene Ontology Categories:
