siRNA Details
Gene Symbol: TMPRSS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042575.2 | NP_001036040.2 | 7 | 963 | ||
NR_026734.1 | 8 | 1076 | ||||
XM_011512754.1 | XP_011511056.1 | 8 | 1408 | |||
XM_011512758.1 | XP_011511060.1 | 7 | 829 | |||
GenBank mRNA | AK131211.1 | - | 8 | 1076 | ||
BC117322.1 | - | 7 | 875 | 40125873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
