siRNA Details
Gene Symbol: TMPRSS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042575.2 | NP_001036040.2 | 16 | 2246 | ||
NR_026734.1 | 17 | 2359 | ||||
XM_011512754.1 | XP_011511056.1 | 17 | 2691 | |||
XM_011512758.1 | XP_011511060.1 | 16 | 2112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
