siRNA Details
Gene Symbol: CFAP157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012502.2 | NP_001012520.2 | 9 | 1616 | ||
XM_006717064.3 | XP_006717127.2 | 8 | 2157 | |||
XM_017014630.1 | XP_016870119.1 | 9 | 1683 | |||
GenBank mRNA | AK094948.1 | - | 8 | 2271 | ||
AL833241.1 | - | 2 | 624 | |||
BC141809.1 | - | 9 | 1693 | 40147564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
