siRNA Details
Gene Symbol: CFAP157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012502.2 | NP_001012520.2 | 3 | 587 | ||
XM_006717064.3 | XP_006717127.2 | 3 | 1315 | |||
XM_011518559.2 | XP_011516861.2 | 3 | 1318 | |||
XM_017014629.1 | XP_016870118.1 | 3 | 1315 | |||
XM_017014630.1 | XP_016870119.1 | 3 | 654 | |||
GenBank mRNA | AK094948.1 | - | 3 | 511 | ||
AK126437.1 | - | 1 | 16 | |||
BC141809.1 | - | 3 | 560 | 40147564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
