siRNA Details
Gene Symbol: FLJ13224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026806.1 | 1 | 1003 | |||
GenBank mRNA | AK023286.1 | - | 1 | 1003 | ||
BC118557.1 | - | 1 | 830 | |||
BC118637.1 | - | 1 | 830 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
