siRNA Details
Gene Symbol: MSANTD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318747.1 | NP_001305676.1 | 3 | 1202 | ||
NM_001318748.1 | NP_001305677.1 | 3 | 1198 | |||
NM_001318749.1 | NP_001305678.1 | 3 | 916 | |||
NM_001318750.1 | NP_001305679.1 | 3 | 843 | |||
NM_032424.2 | NP_115800.1 | 3 | 2076 | |||
XM_011543022.2 | XP_011541324.1 | 4 | 2104 | |||
XM_017018416.1 | XP_016873905.1 | 4 | 1214 | |||
XM_017018417.1 | XP_016873906.1 | 3 | 801 | |||
GenBank mRNA | AB058729.1 | - | 3 | 2016 | ||
AF253976.1 | - | 2 | 266 | |||
AK027790.1 | - | 2 | 236 | |||
AK293050.1 | - | 3 | 872 | |||
AL834523.1 | - | 3 | 766 | |||
BC041118.1 | - | 3 | 773 | 5526792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
