siRNA Details
Gene Symbol: EIF3J-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034170.1 | 2 | 1702 | |||
NR_034171.1 | 3 | 1237 | ||||
GenBank mRNA | AK092547.1 | - | 2 | 1452 | ||
BC066970.1 | - | 2 | 1702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
