siRNA Details
Gene Symbol: ZBTB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137601.2 | NP_001131073.1 | 1 | 1172 | ||
XM_017020911.1 | XP_016876400.1 | 2 | 1276 | |||
GenBank mRNA | AK125003.1 | - | 1 | 288 | ||
AX721091.1 | - | 2 | 1277 | |||
BC157833.1 | - | 1 | 1182 | 9057052 | ||
BC171822.1 | - | 1 | 1182 | 9054476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
