siRNA Details
Gene Symbol: GBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002188.3 | 11 | 1198 | |||
GenBank mRNA | AK295259.1 | - | 8 | 1143 | ||
AK297653.1 | - | 8 | 1118 | |||
BC000349.2 | - | 9 | 1035 | |||
BC030240.1 | - | 11 | 1077 | |||
KJ690775.1 | - | 9 | 951 |
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KJ690772.1 | - | 10 | 1200 | ||
KJ690773.1 | - | 9 | 1310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
