siRNA Details
Gene Symbol: GBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002188.3 | 4 | 395 | |||
GenBank mRNA | AK295259.1 | - | 3 | 305 | ||
AK297653.1 | - | 3 | 280 | |||
BC000349.2 | - | 3 | 307 | |||
BC030240.1 | - | 4 | 274 | |||
BC092482.1 | - | 3 | 305 | |||
KJ690774.1 | - | 3 | 223 | |||
KJ690775.1 | - | 3 | 223 | |||
KJ690776.1 | - | 3 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
