siRNA Details
Gene Symbol: ARMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267041.2 | NP_001253970.1 | 21 | 3395 | ||
NM_001267042.2 | NP_001253971.1 | 21 | 3287 | |||
NM_001282342.1 | NP_001269271.1 | 20 | 3318 | |||
NM_015396.5 | NP_056211.2 | 23 | 3564 | |||
XM_006713564.3 | XP_006713627.1 | 22 | 3483 | |||
XM_006713565.2 | XP_006713628.1 | 23 | 3524 | |||
XM_006713566.3 | XP_006713629.1 | 21 | 3378 | |||
XM_011512632.1 | XP_011510934.1 | 22 | 3431 | |||
XM_011512633.1 | XP_011510935.1 | 22 | 3485 | |||
XM_017006095.1 | XP_016861584.1 | 22 | 3285 | |||
XM_017006097.1 | XP_016861586.1 | 21 | 3231 | |||
XM_017006098.1 | XP_016861587.1 | 20 | 3177 | |||
XM_017006099.1 | XP_016861588.1 | 21 | 3268 | |||
GenBank mRNA | AL096748.1 | - | 9 | 2585 | ||
BQ719381.1 | - | 1 | 547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
