siRNA Details
Gene Symbol: ANKRD62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277333.1 | NP_001264262.1 | 11 | 1558 | ||
XM_017025720.1 | XP_016881209.1 | 11 | 1611 | |||
GenBank mRNA | BX649021.1 | - | 6 | 653 | ||
DW010010.1 | - | 7 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
