siRNA Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085399.1 | NP_001078868.1 | 2 | 267 | ||
NM_001085400.1 | NP_001078869.1 | 2 | 267 | |||
GenBank mRNA | AK075468.1 | - | 2 | 266 | ||
BC039540.1 | - | 2 | 265 | 5748847 | ||
DA400382.1 | - | 2 | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
