siRNA Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085399.1 | NP_001078868.1 | 6 | 882 | ||
NM_001085400.1 | NP_001078869.1 | 6 | 882 | |||
XM_017008590.1 | XP_016864079.1 | 4 | 550 | |||
GenBank mRNA | AK075468.1 | - | 6 | 881 | ||
BC039540.1 | - | 6 | 880 | 5748847 | ||
DA491231.1 | - | 2 | 248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
