siRNA Details
Gene Symbol: LEKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193283.1 | NP_001180212.1 | 1 | 493 | ||
XM_005247441.3 | XP_005247498.1 | 1 | 1624 | |||
XM_006713631.3 | XP_006713694.1 | 1 | 565 | |||
XM_011512803.2 | XP_011511105.1 | 1 | 565 | |||
XM_011512804.2 | XP_011511106.1 | 1 | 1624 | |||
XM_011512806.2 | XP_011511108.1 | 1 | 565 | |||
XM_011512807.2 | XP_011511109.1 | 1 | 565 | |||
XM_011512808.2 | XP_011511110.1 | 1 | 565 | |||
XM_011512811.2 | XP_011511113.1 | 1 | 565 | |||
XM_011512815.2 | XP_011511117.1 | 1 | 565 | |||
XM_011512816.2 | XP_011511118.1 | 1 | 565 | |||
GenBank mRNA | AK094480.1 | - | 1 | 566 | ||
BC041967.1 | - | 1 | 493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
