siRNA Details
Gene Symbol: LEKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193283.1 | NP_001180212.1 | 1 | 729 | ||
XM_005247441.3 | XP_005247498.1 | 1 | 1860 | |||
XM_011512804.2 | XP_011511106.1 | 1 | 1860 | |||
GenBank mRNA | AK094480.1 | - | 1 | 802 | ||
BC041967.1 | - | 1 | 729 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
