siRNA Details
Gene Symbol: FAM78B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320302.1 | NP_001307231.1 | 2 | 2463 | ||
NR_135199.1 | 3 | 3395 | ||||
XM_017000417.1 | XP_016855906.1 | 3 | 7601 | |||
XM_017000418.1 | XP_016855907.1 | 3 | 2943 | |||
GenBank mRNA | AB593134.1 | - | 2 | 2463 | ||
AK055347.1 | - | 1 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
