siRNA Details
Gene Symbol: LOC100130872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024569.1 | 4 | 891 | |||
GenBank mRNA | AK091779.1 | - | 1 | 1544 | ||
BC044945.1 | - | 2 | 296 | |||
BC104228.1 | - | 2 | 264 | |||
BC104229.2 | - | 2 | 260 | |||
BC112991.1 | - | 2 | 264 | |||
BC112992.1 | - | 2 | 264 | |||
CR627052.1 | - | 4 | 891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
