siRNA Details
Gene Symbol: LOC100130872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024569.1 | 4 | 4361 | |||
GenBank mRNA | AK091779.1 | - | 2 | 1810 | ||
AK122772.1 | - | 1 | 1413 | |||
BC030552.1 | - | 1 | 1351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
