siRNA Details
Gene Symbol: TCEANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297563.1 | NP_001284492.1 | 5 | 1388 | ||
NM_001297564.1 | NP_001284493.1 | 3 | 1179 | |||
NM_152634.3 | NP_689847.2 | 4 | 1335 | |||
XM_017029316.1 | XP_016884805.1 | 4 | 1292 | |||
XM_017029317.1 | XP_016884806.1 | 4 | 1268 | |||
XM_017029318.1 | XP_016884807.1 | 4 | 1265 | |||
XM_017029319.1 | XP_016884808.1 | 3 | 1221 | |||
GenBank mRNA | AK097032.1 | - | 1 | 768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
