siRNA Details
Gene Symbol: ANXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014279.2 | NP_001014301.1 | 1 | 387 | ||
GenBank mRNA | AY032883.1 | - | 1 | 293 | ||
BC067873.1 | - | 1 | 135 | 30344324 | ||
CN261477.1 | - | 1 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
