siRNA Details
Gene Symbol: NEPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319109.1 | NP_001306038.1 | 2, 3 | 355 | ||
NM_015412.3 | NP_056227.2 | 2, 3 | 330 | |||
NR_134969.1 | 2, 3 | 355 | ||||
XM_005247276.2 | XP_005247333.1 | 2, 3 | 293 | |||
XM_011512635.2 | XP_011510937.1 | 2, 3 | 293 | |||
XM_011512636.2 | XP_011510938.1 | 2, 3 | 293 | |||
XM_017006102.1 | XP_016861591.1 | 2, 3 | 293 | |||
GenBank mRNA | AK024325.1 | - | 2, 3 | 288 | ||
AK293218.1 | - | 2, 3 | 293 | |||
AL117573.1 | - | 2, 3 | 272 | |||
BC014673.2 | - | 2, 3 | 328 | |||
BC067743.1 | - | 2, 3 | 290 | 4812681 | ||
CR749341.1 | - | 2, 3 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
