siRNA Details
Gene Symbol: LOC648987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034085.1 | 1 | 1063 | |||
NR_034086.1 | 1 | 1063 | ||||
NR_104649.1 | 1 | 1063 | ||||
GenBank mRNA | AK097396.1 | - | 1 | 576 | ||
BC013653.1 | - | 1 | 977 | |||
BC034002.1 | - | 1 | 73 | |||
BC039088.1 | - | 1 | 1063 | |||
BC046188.1 | - | 1 | 976 |
Gene Symbol: FLJ32255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104643.1 | 1 | 1513 | |||
GenBank mRNA | AK056817.1 | - | 1 | 1513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
