siRNA Details
Gene Symbol: LOC648987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034086.1 | 2 | 1863 | |||
NR_104649.1 | 3 | 1979 | ||||
NR_104650.1 | 2 | 233 | ||||
NR_104651.1 | 3 | 338 | ||||
GenBank mRNA | BC013653.1 | - | 2 | 1777 | ||
BC039088.1 | - | 3 | 1979 | |||
BF130969.1 | - | 314 | ||||
HY041253.1 | - | 2 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
