siRNA Details
Gene Symbol: LOC648987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034086.1 | 2 | 1999 | |||
NR_104649.1 | 3 | 2115 | ||||
NR_104650.1 | 2 | 369 | ||||
NR_104651.1 | 3 | 474 | ||||
GenBank mRNA | BC013653.1 | - | 2 | 1913 | ||
BC039088.1 | - | 3 | 2115 | |||
BC046188.1 | - | 2 | 1912 | |||
BF130969.1 | - | 450 | ||||
HY041253.1 | - | 2 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
