siRNA Details
Gene Symbol: MEIOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145080.2 | NP_001138552.2 | 6 | 2633 | ||
XM_005257236.3 | XP_005257293.1 | 6 | 2610 | |||
GenBank mRNA | AK097747.1 | - | 1 | 103 | ||
AK302001.1 | - | 4 | 2074 | |||
BC035159.1 | - | 3 | 2325 | 5265358 | ||
BC041481.2 | - | 2 | 662 | |||
BC132822.1 | - | 2 | 1954 | 40146844 | ||
BC136642.1 | - | 2 | 1954 | 9020632 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001127576, CJ046726, BG083749.
Gene Ontology Categories:
