siRNA Details
Gene Symbol: MEIOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145080.2 | NP_001138552.2 | 5 | 2103 | ||
XM_005257236.3 | XP_005257293.1 | 5 | 2080 | |||
GenBank mRNA | AK093167.1 | - | 2 | 1439 | ||
AK302001.1 | - | 3 | 1544 | |||
BC035159.1 | - | 2 | 1795 | 5265358 | ||
BC041481.2 | - | 1 | 132 | |||
BC132822.1 | - | 1 | 1424 | 40146844 | ||
BC136642.1 | - | 1 | 1424 | 9020632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
