siRNA Details
Gene Symbol: TMEM185B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024121.2 | NP_077026.2 | 1 | 1087 | ||
GenBank mRNA | AF530474.1 | - | 1 | 1087 | ||
AK289880.1 | - | 1 | 1049 | |||
BC080607.1 | - | 1 | 1054 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
