siRNA Details
Gene Symbol: NEURL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142651.2 | NP_001136123.1 | 5 | 5372 | ||
NM_001308177.1 | NP_001295106.1 | 4 | 4826 | |||
NM_001308178.1 | NP_001295107.1 | 4 | 4652 | |||
GenBank mRNA | AK026748.1 | - | 1 | 1165 | ||
AK091605.1 | - | 1 | 1869 | |||
AK096661.1 | - | 1 | 2959 | |||
AK126311.1 | - | 2 | 2411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
