siRNA Details
Gene Symbol: FAM215A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026770.1 | 1 | 588 | |||
GenBank mRNA | AF143236.1 | - | 1 | 601 | ||
BC069097.1 | - | 1 | 588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
