siRNA Details
Gene Symbol: ZNF594 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032530.1 | NP_115919.1 | 2 | 305 | ||
XM_005256827.2 | XP_005256884.1 | 2 | 618 | |||
GenBank mRNA | AB058774.2 | - | 1 | 290 | ||
AK131271.1 | - | 2 | 305 | |||
BC136604.1 | - | 1 | 283 | 9020594 | ||
BX472376.1 | - | 2 | 301 | |||
DA448705.1 | - | 2 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
