siRNA Details
Gene Symbol: TIMM23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290117.1 | NP_001277046.1 | 7 | 816 | ||
NM_001290118.1 | NP_001277047.1 | 5 | 672 | |||
NR_110767.1 | 8 | 940 | ||||
GenBank mRNA | AK098044.1 | - | 8 | 925 | ||
AK293228.1 | - | 5 | 624 | |||
AK295227.1 | - | 7 | 1126 | |||
AK295370.1 | - | 7 | 768 | |||
AK316490.1 | - | 7 | 768 | |||
AK316549.1 | - | 7 | 817 | |||
BC035067.1 | - | 2 | 694 |
Gene Symbol: PARGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF130105.1 | - | 1 | 1726 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
