siRNA Details
Gene Symbol: YOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276320.1 | NP_001263249.1 | 4 | 2537 | ||
NM_018566.3 | NP_061036.3 | 2 | 2511 | |||
GenBank mRNA | AL833081.2 | - | 2 | 2444 | ||
BC137166.1 | - | 2 | 2503 | 9021163 | ||
BC137167.1 | - | 2 | 2503 | 9021164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
