siRNA Details
Gene Symbol: FBXO43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029860.3 | NP_001025031.2 | 2 | 1539 | ||
NR_036491.1 | 2 | 1583 | ||||
XM_011516987.2 | XP_011515289.1 | 2 | 1485 | |||
XM_011516989.2 | XP_011515291.1 | 2 | 1529 | |||
XM_011516990.1 | XP_011515292.1 | 2 | 755 | |||
XM_011516992.2 | XP_011515294.1 | 2 | 1485 | |||
XM_017013358.1 | XP_016868847.1 | 2 | 1485 | |||
GenBank mRNA | AK310518.1 | - | 2 | 1109 | ||
BC090892.1 | - | 2 | 1512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
