siRNA Details
Gene Symbol: LYSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286812.1 | NP_001273741.1 | 2 | 327 | ||
NM_198273.2 | NP_938014.1 | 2 | 327 | |||
XM_005248421.4 | XP_005248478.1 | 2 | 792 | |||
GenBank mRNA | AK291478.1 | - | 2 | 324 | ||
AL832721.1 | - | 2 | 308 | |||
BC058027.1 | - | 2 | 327 | |||
BC136740.1 | - | 2 | 219 | 9020730 | ||
BC136741.1 | - | 2 | 219 | 9020731 | ||
BP873090.1 | - | 2 | 328 | |||
BX500166.1 | - | 2 | 336 | |||
BX537972.1 | - | 2 | 297 | |||
CR933624.1 | - | 3 | 388 | |||
DN994993.1 | - | 2 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
