siRNA Details
Gene Symbol: RPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122965.1 | NP_001116437.1 | 3 | 1891 | ||
GenBank mRNA | AK096436.1 | - | 1 | 801 | ||
AY396742.1 | - | 3 | 1891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
