siRNA Details
Gene Symbol: TDRD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010870.2 | NP_001010870.1 | 1 | 4670 | ||
NM_001168359.1 | NP_001161831.1 | 1 | 4670 | |||
GenBank mRNA | AF039442.1 | - | 1 | 607 | ||
AK125838.1 | - | 1 | 1758 | |||
BX648686.1 | - | 1 | 1202 | |||
CR936792.1 | - | 1 | 3020 | |||
EF185284.1 | - | 1 | 1155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
